[4-(4-Methoxyphenoxy)phenyl]methanamine 95%

Lieferant: Apollo Scientific

OR32439-1G
APOSOR32439-1GEA 387 EUR
APOSOR32439-1G
[4-(4-Methoxyphenoxy)phenyl]methanamine 95%
[4-(4-Methoxyphenoxy)phenyl]methanamine
Formel: C₁₄H₁₅NO₂
Molecular Weight: 229,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01814768
CAS-Nummer: 477868-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR