2-[(Prop-2-en-1-yl)amino]-3,4-dihydroquinazolin-4-one, tech
Lieferant: Apollo Scientific
OR32441-1G
APOSOR32441-1GEA
1570
EUR
APOSOR32441-1G
2-[(Prop-2-en-1-yl)amino]-3,4-dihydroquinazolin-4-one, tech
2-[(Prop-2-en-1-yl)amino]-3,4-dihydroquinazolin-4-one
Formel:
C₁₁H₁₁N₃O Molecular Weight: 201,225 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01814694 CAS-Nummer: 212143-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...