5,6-Dibromocyclohex-2-ene-1,4-diol, tech
Lieferant: Apollo Scientific
OR32443-1G
APOSOR32443-1GEA
1570
EUR
APOSOR32443-1G
5,6-Dibromocyclohex-2-ene-1,4-diol, tech
5,6-Dibromocyclohex-2-ene-1,4-diol
Formel:
C₆H₈Br₂O₂ Molecular Weight: 271,935 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01568911 CAS-Nummer: 317821-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...