2-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)acetonitrile, tech

Lieferant: Apollo Scientific

OR32444-1G
APOSOR32444-1GEA 671 EUR
APOSOR32444-1G
2-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)acetonitrile, tech
2-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)acetonitrile
Formel: C₄H₂Br₂N₄
Molecular Weight: 265,893 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01814608
CAS-Nummer: 477869-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR