(2Z)-2-Cyano-N-(3,5-dichlorophenyl)-3-(dimethylamino)prop-2-enamide, tech
Lieferant: Apollo Scientific
OR32448-1G
APOSOR32448-1GEA
1570
EUR
APOSOR32448-1G
(2Z)-2-Cyano-N-(3,5-dichlorophenyl)-3-(dimethylamino)prop-2-enamide, tech
(2Z)-2-Cyano-N-(3,5-dichlorophenyl)-3-(dimethylamino)prop-2-enamide
Formel:
C₁₂H₁₁Cl₂N₃O Molecular Weight: 284,141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02102377 CAS-Nummer: 477870-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...