Methyl (2E)-3-(quinolin-3-yl)prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32450-1G
APOSOR32450-1GEA 1570 EUR
APOSOR32450-1G
Methyl (2E)-3-(quinolin-3-yl)prop-2-enoate, tech
Methyl (2E)-3-(quinolin-3-yl)prop-2-enoate
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02068445
CAS-Nummer: 66417-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR