4-(3-Chlorophenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine 95%
Lieferant: Apollo Scientific
OR32457-1G
APOSOR32457-1GEA
874
EUR
APOSOR32457-1G
4-(3-Chlorophenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine 95%
4-(3-Chlorophenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine
Formel:
C₁₂H₁₂ClN₃ Molecular Weight: 233.7 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02102424 CAS-Nummer: 18094-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...