1-Butyl-6-oxo-2-phenylpiperidine-3-carboxylic acid, tech

Lieferant: Apollo Scientific

OR32459-1G
APOSOR32459-1GEA 1570 EUR
APOSOR32459-1G
1-Butyl-6-oxo-2-phenylpiperidine-3-carboxylic acid, tech
1-Butyl-6-oxo-2-phenylpiperidine-3-carboxylic acid
Formel: C₁₆H₂₁NO₃
Molecular Weight: 275,343 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571318
CAS-Nummer: 164933-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR