Ethyl 2-(4-methyl-3-oxo-3,4-dihydroquinoxalin-2-yl)acetate, tech

Lieferant: Apollo Scientific

OR32460-1G
APOSOR32460-1GEA 1570 EUR
APOSOR32460-1G
Ethyl 2-(4-methyl-3-oxo-3,4-dihydroquinoxalin-2-yl)acetate, tech
Ethyl 2-(4-methyl-3-oxo-3,4-dihydroquinoxalin-2-yl)acetate
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02185980
CAS-Nummer: 14152-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR