N-(4-Chlorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide, tech

Lieferant: Apollo Scientific

OR32461-1G
APOSOR32461-1GEA 0 EUR
APOSOR32461-1G
N-(4-Chlorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide, tech
N-(4-Chlorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide
Formel: C₁₃H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02102447
CAS-Nummer: 477871-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR