2-(1-Methyl-1H-pyrrol-2-yl)-2-oxo-N-phenylacetamide, tech

Lieferant: Apollo Scientific

OR32462-1G
APOSOR32462-1GEA 1570 EUR
APOSOR32462-1G
2-(1-Methyl-1H-pyrrol-2-yl)-2-oxo-N-phenylacetamide, tech
2-(1-Methyl-1H-pyrrol-2-yl)-2-oxo-N-phenylacetamide
Formel: C₁₃H₁₂N₂O₂
Molecular Weight: 228,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102448
CAS-Nummer: 477871-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR