8-Benzyl-2,8-diazaspiro[4.5]decane-1,3-dione 95%

Lieferant: Apollo Scientific

OR32466-1G
APOSOR32466-1GEA 671 EUR
APOSOR32466-1G
8-Benzyl-2,8-diazaspiro[4.5]decane-1,3-dione 95%
8-Benzyl-2,8-diazaspiro[4.5]decane-1,3-dione
Formel: C₁₅H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11043250
CAS-Nummer: 1463-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR