6-(3-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech

Lieferant: Apollo Scientific

OR32467-1G
APOSOR32467-1GEA 1570 EUR
APOSOR32467-1G
6-(3-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
6-(3-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine
Formel: C₁₃H₁₁ClN₄
Molecular Weight: 258,706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102690
CAS-Nummer: 85841-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR