1,5-Dimethyl-4-phenylimidazolidin-2-one 95%

Lieferant: Apollo Scientific

OR32468-1G
APOSOR32468-1GEA 874 EUR
APOSOR32468-1G
1,5-Dimethyl-4-phenylimidazolidin-2-one 95%
1,5-Dimethyl-4-phenylimidazolidin-2-one
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01456559
CAS-Nummer: 103774-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR