2-[(Furan-2-yl)methylidene]-2,3-dihydro-1H-indene-1,3-dione, tech

Lieferant: Apollo Scientific

OR32469-1G
APOSOR32469-1GEA 1570 EUR
APOSOR32469-1G
2-[(Furan-2-yl)methylidene]-2,3-dihydro-1H-indene-1,3-dione, tech
2-[(Furan-2-yl)methylidene]-2,3-dihydro-1H-indene-1,3-dione
Formel: C₁₄H₈O₃
Molecular Weight: 224,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00219001
CAS-Nummer: 29874-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR