(2E)-1-(1-Benzothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32471-1G
APOSOR32471-1GEA 1570 EUR
APOSOR32471-1G
(2E)-1-(1-Benzothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-(1-Benzothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₃H₁₃NOS
Molecular Weight: 231,313 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617977
CAS-Nummer: 893441-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR