1-(5-Chlorothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32474-1G
APOSOR32474-1GEA 1570 EUR
APOSOR32474-1G
1-(5-Chlorothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one, tech
1-(5-Chlorothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one
Formel: C₁₀H₁₂ClNOS
Molecular Weight: 229,726 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617982
CAS-Nummer: 860788-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR