5-Methyl-2-phenylpyrazolo[1,5-a]pyrimidin-7-ol 95%

Lieferant: Apollo Scientific

OR32479-1G
APOSOR32479-1GEA 671 EUR
APOSOR32479-1G
5-Methyl-2-phenylpyrazolo[1,5-a]pyrimidin-7-ol 95%
5-Methyl-2-phenylpyrazolo[1,5-a]pyrimidin-7-ol
Formel: C₁₃H₁₁N₃O
Molecular Weight: 225,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00634653
CAS-Nummer: 90019-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR