Cyanomethyl 4-(1H-pyrrol-1-yl)benzoate, tech
Lieferant: Apollo Scientific
OR32481-1G
APOSOR32481-1GEA
1570
EUR
APOSOR32481-1G
Cyanomethyl 4-(1H-pyrrol-1-yl)benzoate, tech
Cyanomethyl 4-(1H-pyrrol-1-yl)benzoate
Formel:
C₁₃H₁₀N₂O₂ Molecular Weight: 226,231 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03012862 CAS-Nummer: 439095-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...