N-[4-(2-Phenylethynyl)phenyl]methanesulphonamide, tech

Lieferant: Apollo Scientific

OR32482-1G
APOSOR32482-1GEA 1570 EUR
APOSOR32482-1G
N-[4-(2-Phenylethynyl)phenyl]methanesulphonamide, tech
N-[4-(2-Phenylethynyl)phenyl]methanesulphonamide
Formel: C₁₅H₁₃NO₂S
Molecular Weight: 271,334 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012868
CAS-Nummer: 439095-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR