3,5-Dichlorphenylhydraziniumchlorid 98%

Lieferant: Apollo Scientific
Achtung

OR3249-100G
APOSOR3249-100GEA 0 EUR
APOSOR3249-100G
3,5-Dichlorphenylhydraziniumchlorid 98%
3,5-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00012938
CAS-Nummer: 63352-99-8
EINECS: 264-109-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR