6H,7H,8H,9H,10H,12H-Azepino[2,1-b]quinazolin-12-one 95%

Lieferant: Apollo Scientific

OR32495-1G
APOSOR32495-1GEA 671 EUR
APOSOR32495-1G
6H,7H,8H,9H,10H,12H-Azepino[2,1-b]quinazolin-12-one 95%
6H,7H,8H,9H,10H,12H-Azepino[2,1-b]quinazolin-12-one
Formel: C₁₃H₁₄N₂O
Molecular Weight: 214,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00661748
CAS-Nummer: 4425-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR