2-(1H-Pyrrol-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione, tech

Lieferant: Apollo Scientific

Synonyme: 2-(1H-Pyrrol-1-yl)isoindoline-1,3-dione

OR32498-1G
APOSOR32498-1GEA 1410 EUR
APOSOR32498-1G
2-(1H-Pyrrol-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione, tech
2-(1H-Pyrrol-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Formel: C₁₂H₈N₂O₂
Molecular Weight: 212,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00837647
CAS-Nummer: 885-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR