Ethyl-4-(4-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-(4-aminophenyl)piperazine-1-carboxylate

OR32500-1G
APOSOR32500-1GEA 538 EUR
APOSOR32500-1G
Ethyl-4-(4-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 95%
Ethyl-4-(4-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₁₃H₁₉N₃O₂
Molecular Weight: 249,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848963
CAS-Nummer: 16154-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR