2-(4-Benzylpiperazino)-2-methylpropanal 95%
Lieferant: Apollo Scientific
OR32501-1G
APOSOR32501-1GEA
380
EUR
APOSOR32501-1G
2-(4-Benzylpiperazino)-2-methylpropanal 95%
2-(4-Benzylpiperazino)-2-methylpropanal
Formel:
C₁₅H₂₂N₂O Molecular Weight: 246.36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663785 CAS-Nummer: 886360-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...