4-[(6-Chloropyridin-3-yl)methoxy]benzonitrile, tech
Lieferant: Apollo Scientific
OR32504-1G
APOSOR32504-1GEA
1570
EUR
APOSOR32504-1G
4-[(6-Chloropyridin-3-yl)methoxy]benzonitrile, tech
4-[(6-Chloropyridin-3-yl)methoxy]benzonitrile
Formel:
C₁₃H₉ClN₂O Molecular Weight: 244,676 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124800 CAS-Nummer: 861207-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...