2-[(E)-[(3-Chloro-4-methoxyphenyl)methylidene]amino]ethan-1-ol, tech

Lieferant: Apollo Scientific

OR32508-1G
APOSOR32508-1GEA 1570 EUR
APOSOR32508-1G
2-[(E)-[(3-Chloro-4-methoxyphenyl)methylidene]amino]ethan-1-ol, tech
2-[(E)-[(3-Chloro-4-methoxyphenyl)methylidene]amino]ethan-1-ol
Formel: C₁₀H₁₂ClNO₂
Molecular Weight: 213,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124828
CAS-Nummer: 861207-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR