1-[(6-Chloropyridin-3-yl)methyl]-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32509-1G
APOSOR32509-1GEA 1570 EUR
APOSOR32509-1G
1-[(6-Chloropyridin-3-yl)methyl]-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
1-[(6-Chloropyridin-3-yl)methyl]-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₄H₁₂ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD05668747
CAS-Nummer: 861207-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR