N'-Cyano-4-pyridinecarboximidamide

Lieferant: Apollo Scientific
Achtung

OR3251-1G
APOSOR3251-1GEA 105 EUR
APOSOR3251-1G
N'-Cyano-4-pyridinecarboximidamide
N'-Cyano-4-pyridinecarboximidamide
Formel: C₇H₆N₄
Molecular Weight: 146,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11553048
CAS-Nummer: 23275-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR