6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32510-1G
APOSOR32510-1GEA 1570 EUR
APOSOR32510-1G
6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech
6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-3-carbonitrile
Formel: C₁₂H₁₁N₃
Molecular Weight: 197,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124853
CAS-Nummer: 861207-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR