2-(4-Benzylpiperazino)nicotinaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Benzylpiperazin-1-yl)pyridine-3-carbaldehyde

OR32512-1G
APOSOR32512-1GEA 671 EUR
APOSOR32512-1G
2-(4-Benzylpiperazino)nicotinaldehyde 95%
2-(4-Benzylpiperazino)nicotinaldehyde
Formel: C₁₇H₁₉N₃O
Molecular Weight: 281,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659545
CAS-Nummer: 885949-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR