2-(4-Methoxyphenyl)nicotinaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Methoxyphenyl)pyridine-3-carbaldehyde

OR32513-1G
APOSOR32513-1GEA 671 EUR
APOSOR32513-1G
2-(4-Methoxyphenyl)nicotinaldehyde 95%
2-(4-Methoxyphenyl)nicotinaldehyde
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659546
CAS-Nummer: 885949-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR