1-Acetyl-N-(3-ethynylphenyl)piperidine-4-carboxamide, tech

Lieferant: Apollo Scientific

OR32514-1G
APOSOR32514-1GEA 1570 EUR
APOSOR32514-1G
1-Acetyl-N-(3-ethynylphenyl)piperidine-4-carboxamide, tech
1-Acetyl-N-(3-ethynylphenyl)piperidine-4-carboxamide
Formel: C₁₆H₁₈N₂O₂
Molecular Weight: 270,326 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496153
CAS-Nummer: 861208-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR