3,6-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid, tech

Lieferant: Apollo Scientific

OR32515-1G
APOSOR32515-1GEA 1570 EUR
APOSOR32515-1G
3,6-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid, tech
3,6-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₄Cl₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08064716
CAS-Nummer: 923249-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR