N-[(4-Bromophenyl)methyl]acetamide 97%

Lieferant: Apollo Scientific

Synonyme: N-(4-Bromobenzyl)acetamide

OR32516-1G
APOSOR32516-1GEA 335 EUR
APOSOR32516-1G
N-[(4-Bromophenyl)methyl]acetamide 97%
N-[(4-Bromophenyl)methyl]acetamide
Formel: C₉H₁₀BrNO
Molecular Weight: 228.086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14659390
CAS-Nummer: 90561-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR