Methyl 2-[(3-methylphenyl)formamido]acetate, tech

Lieferant: Apollo Scientific

OR32518-1G
APOSOR32518-1GEA 1570 EUR
APOSOR32518-1G
Methyl 2-[(3-methylphenyl)formamido]acetate, tech
Methyl 2-[(3-methylphenyl)formamido]acetate
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01213551
CAS-Nummer: 1208-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR