1-(2-Chloroethyl)-3,5-dimethyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR32524-1G
APOSOR32524-1GEA 339 EUR
APOSOR32524-1G
1-(2-Chloroethyl)-3,5-dimethyl-1H-pyrazole 95%
1-(2-Chloroethyl)-3,5-dimethyl-1H-pyrazole
Formel: C₇H₁₁ClN₂
Molecular Weight: 158.629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215355
CAS-Nummer: 38460-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR