1-[(2,6-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech

Lieferant: Apollo Scientific

OR32526-1G
APOSOR32526-1GEA 1570 EUR
APOSOR32526-1G
1-[(2,6-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech
1-[(2,6-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea
Formel: C₁₁H₁₃Cl₂N₃O₂
Molecular Weight: 290,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139682
CAS-Nummer: 338395-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR