Ethyl (2E)-3-amino-3-(4-phenylpiperazin-1-yl)prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32529-1G
APOSOR32529-1GEA 1570 EUR
APOSOR32529-1G
Ethyl (2E)-3-amino-3-(4-phenylpiperazin-1-yl)prop-2-enoate, tech
Ethyl (2E)-3-amino-3-(4-phenylpiperazin-1-yl)prop-2-enoate
Formel: C₁₅H₂₁N₃O₂
Molecular Weight: 275,346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664733
CAS-Nummer: 79823-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR