N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-methoxy-N,N-dimethylpyrimidin-2-amine

OR32532-1G
APOSOR32532-1GEA 488 EUR
APOSOR32532-1G
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine 95%
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine
Formel: C₇H₁₀ClN₃O
Molecular Weight: 187,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935946
CAS-Nummer: 13012-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR