3-Hydroxy-2-methyl-3-(2-phenylethynyl)-2,3-dihydro-1H-isoindol-1-one, tech

Lieferant: Apollo Scientific

OR32535-1G
APOSOR32535-1GEA 1570 EUR
APOSOR32535-1G
3-Hydroxy-2-methyl-3-(2-phenylethynyl)-2,3-dihydro-1H-isoindol-1-one, tech
3-Hydroxy-2-methyl-3-(2-phenylethynyl)-2,3-dihydro-1H-isoindol-1-one
Formel: C₁₇H₁₃NO₂
Molecular Weight: 263,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215773
CAS-Nummer: 143647-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR