5-(4-Chlorophenyl)-2-[(dimethylamino)methylidene]cyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR32536-1G
APOSOR32536-1GEA 1570 EUR
APOSOR32536-1G
5-(4-Chlorophenyl)-2-[(dimethylamino)methylidene]cyclohexane-1,3-dione, tech
5-(4-Chlorophenyl)-2-[(dimethylamino)methylidene]cyclohexane-1,3-dione
Formel: C₁₅H₁₆ClNO₂
Molecular Weight: 277,746 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172130
CAS-Nummer: 338399-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR