1,1-Diphenyl-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol, tech

Lieferant: Apollo Scientific

OR32539-1G
APOSOR32539-1GEA 0 EUR
APOSOR32539-1G
1,1-Diphenyl-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol, tech
1,1-Diphenyl-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol
Formel: C₁₆H₁₅N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04124868
CAS-Nummer: 76674-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR