4,6-Dimethyl-2-pyrrolidin-1-ylnicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR3254-1G
APOSOR3254-1GEA 81.2 EUR
APOSOR3254-1G
4,6-Dimethyl-2-pyrrolidin-1-ylnicotinonitrile
4,6-Dimethyl-2-pyrrolidin-1-ylnicotinonitrile
Formel: C₁₂H₁₅N₃
Molecular Weight: 201,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04209734
CAS-Nummer: 693254-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR