2-(Morpholin-4-yl)-1,1-diphenylethan-1-ol, tech

Lieferant: Apollo Scientific

OR32540-1G
APOSOR32540-1GEA 1570 EUR
APOSOR32540-1G
2-(Morpholin-4-yl)-1,1-diphenylethan-1-ol, tech
2-(Morpholin-4-yl)-1,1-diphenylethan-1-ol
Formel: C₁₈H₂₁NO₂
Molecular Weight: 283,365 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231484
CAS-Nummer: 13150-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR