4-Methoxy-2-(4-phenylpiperazin-1-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32542-1G
APOSOR32542-1GEA 1570 EUR
APOSOR32542-1G
4-Methoxy-2-(4-phenylpiperazin-1-yl)pyridine-3-carbonitrile, tech
4-Methoxy-2-(4-phenylpiperazin-1-yl)pyridine-3-carbonitrile
Formel: C₁₇H₁₈N₄O
Molecular Weight: 294,351 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975443
CAS-Nummer: 338773-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR