Ethyl 2-(2-oxo-1,2-dihydroquinoxalin-1-yl)acetate, tech

Lieferant: Apollo Scientific

OR32543-1G
APOSOR32543-1GEA 1570 EUR
APOSOR32543-1G
Ethyl 2-(2-oxo-1,2-dihydroquinoxalin-1-yl)acetate, tech
Ethyl 2-(2-oxo-1,2-dihydroquinoxalin-1-yl)acetate
Formel: C₁₂H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05155084
CAS-Nummer: 154640-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR