6-Methyl-2-(pyridin-4-yl)pyrimidin-4-ol 95%

Lieferant: Apollo Scientific

OR32547-1G
APOSOR32547-1GEA 71.2 EUR
APOSOR32547-1G
6-Methyl-2-(pyridin-4-yl)pyrimidin-4-ol 95%
6-Methyl-2-(pyridin-4-yl)pyrimidin-4-ol
Formel: C₁₀H₉N₃O
Molecular Weight: 187,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00106896
CAS-Nummer: 59341-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR