3-Acetyl-4-methoxy-1,2-dihydroquinolin-2-one, tech

Lieferant: Apollo Scientific

OR32550-1G
APOSOR32550-1GEA 1570 EUR
APOSOR32550-1G
3-Acetyl-4-methoxy-1,2-dihydroquinolin-2-one, tech
3-Acetyl-4-methoxy-1,2-dihydroquinolin-2-one
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316892
CAS-Nummer: 400079-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR