1H,2H-Naphtho[2,1-b]furan-1-one, tech
Lieferant: Apollo Scientific
OR32551-1G
APOSOR32551-1GEA
1570
EUR
APOSOR32551-1G
1H,2H-Naphtho[2,1-b]furan-1-one, tech
1H,2H-Naphtho[2,1-b]furan-1-one
Formel:
C₁₂H₈O₂ Molecular Weight: 184,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975011 CAS-Nummer: 19997-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...